Learn More
Thermo Scientific™ National Flat-Bottom Vial Inserts
Shop All Thermo Scientific Products
Click to view available options
Unit Size:
Pack of 1000
Description
Thermo Scientific™ National Flat-Bottom Vial Inserts are bulk-packed for convenience.
Specifications
Specifications
Description | 200μL; 5mm OD |
Diameter (Metric) Outer | 5 mm |
Volume (Metric) | 200 μL |
Type | Insert |