Learn More
NFKBIL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26273-1-AP-150UL
 View more versions of this product
Product Code. 16854785
  • 3058.99 DKK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
NF-kappa-B inhibitor-like protein 1 (NFKBIL1), also named Ikb like and IkbL1, is a 381 amino acid member of the inhibitor of NF-kappaB (IkappaB) family of proteins. NFKBIL1 contains two ANK repeats and is expressed in the rheumatoid synovial lining and sub-lining layers and co-localized in CD68+ and CD3+, but not Factor VIII+ cells. It is reported that defects in NFKBIL1 expression causes of susceptibility to rheumatoid arthritis (RA), which is a systemic inflammatory disease with autoimmune features and affects the joints. Rheumatoid arthritis (RA) is characterized by inflammatory changes in the synovial membranes and articular structures, widespread fibrinoid degeneration of the collagen fibers in mesenchymal tissues, and by atrophy and rarefaction of bony structures.
Specifications
Specifications
NFKBIL1
Polyclonal
Unconjugated
NFKBIL1
I kappa B like protein, IkappaBL, IKBL, LST1, NFKBIL, NFKBIL1
Rabbit
Antigen Affinity Chromatography
RUO
4795
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UBC1
NFKBIL1
NFKBIL1 Fusion Protein Ag21978
150 μL
Primary
Human
Antibody
IgG