Learn More
Nickel Aluminum, Thermo Scientific Chemicals
Product Code. 11359147
Click to view available options
Quantity:
100 g
500 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Wear resistant coatings
Nickel Aluminum is used as wear resistant coatings. It is also used for the preparation of Raney™ Nickel catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 12635-27-7
Molecular Formula AlNi
Molecular Weight (g/mol) 85.675
MDL Number MFCD00167392
InChI Key NPXOKRUENSOPAO-UHFFFAOYSA-N
Synonym Aluminum Nickel
PubChem CID 6336846
IUPAC Name aluminum;nickel
SMILES [Al].[Ni]
Specifications
Color Black to Gray
Avg. Mol. Wt. or Mol. Wt. Range Ni:Al; 50:50 wt%
UN Number UN3208
Quantity 500 g
Solubility Information Insoluble in water.
Odor Odorless
Sensitivity Water reactive
Form Powder
Chemical Name or Material Nickel Aluminum