Learn More
Ansell BioClean™ Biotac BIOTAC Nitrile Gloves
Latex-free, powder free cleanroom gloves are flexible and resistant to a wide range of chemicals.
Brand: Ansell BIOTAC-M
1816.55 DKK valid until 2025-12-31
Use promo code "24381" to get your promotional price.
This item is not returnable.
View return policy
Description
- 12 in. length
- Ambidextrous
- AQL 1.5
- Finger-textured, powder free
- Nitrile is resistant to a wide range of chemicals
- ESD properties
- Processed to ensure ISO Class 5 Compatibility
- Suitable for Production and Manufacturing, Controlled and Critical Environments, Lab and R&D
Specifications
CE 0598, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 455 Part 2, Food Contact, Category III | |
1.5 | |
Partially Textured | |
0.17 mm | |
Yes | |
4.3 mil | |
Powder Free | |
Non-sterile | |
0.17 mm | |
BIOTAC-M | |
12 in. |
Class 100 (ISO 5) | |
White | |
6.7 mil | |
Ambidextrous | |
Nitrile | |
0.11 mm | |
Medium | |
6.7 mil | |
ISO 9001, PPE Regulation 2016 425 Module D | |
BioClean™ Biotac BIOTAC | |
30.5 cm |