Learn More
NLGN3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29319-1-AP-20UL
Description
The NLGN3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NLGN3. This antibody recognizes human, mouse, rat antigen. The NLGN3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
NLGN3 | |
600 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat | |
150 kDa | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
ASPGX1, AUTSX1, Gliotactin homolog, HNL3, KIAA1480, neuroligin 3, NL3, NLGN3 | |
20 μL | |
54413 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |