Learn More
Brady™ Non-Conductive Nylon Cable
158.51 DKK - 181.00 DKK
Specifications
Product Type Nylon Cable
Color Red
Diameter (Metric) 3.18 mm
Quantity 1 Ea.
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
11771855
View Documents
Brady™
050945
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11791855
View Documents
Brady™
050948
3.65 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications
Nylon Cable
Red
3.18 mm
1 Ea.