Learn More
Macherey-Nagel™ NucleoSpin™ DNA Forensic, Mini kit for DNA from forensic traces
Spin technology based isolation of genomic DNA from forensic samples (e.g., casework samples, contact traces). Complete kits (spin columns, buffers, collection tubes, liquid proteinase K, reducing agent TCEP) available in several quantities.
417.00 DKK - 7005.00 DKK
Specifications
Concentration | 10 to 30 ng/μL |
---|---|
Content And Storage | Room temperature (18-25 °C) |
Format | Mini Spin Column |
Isolation Technology | Silica Membrane Technology |
Sample Type | Casework Samples, Contact Traces (e.g., Dried Blood Spots, Cigarette Filters, Swabs) |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15777307
|
Macherey-Nagel™
740840.10 |
10 Preps |
417.00 DKK
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15797307
|
Macherey-Nagel™
740840.50 |
50 Preps |
1540.00 DKK
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15787307
|
Macherey-Nagel™
740840.250 |
250 Preps |
7005.00 DKK
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NucleoSpin™ DNA Forensic Kits
- Excellent genomic DNA purity from all casework samples
- Procedure is based on reversible adsorption of nucleic acids to the silica membrane
- Sample sources include buccal swabs or small tissue/cell samples (dried blood spots, trace and touch-samples or cigarette filters)
- Uniform buffer system for single sample analysis and for high throughput analysis
- Ethylene oxide-treated plastic materials
- Conformity to ISO 18385 for doubtless DNA profiling
- Applications: STR analysis, qPCR, any kind of enzymatic reaction
Specifications
10 to 30 ng/μL | |
Mini Spin Column | |
Casework Samples, Contact Traces (e.g., Dried Blood Spots, Cigarette Filters, Swabs) | |
Elution volume: 50-100 μL | |
Vortex mixer | |
NucleoSpin DNA Forensic columns, buffers, collection tubes (2 mL), liquid proteinase K, reducing agent TCEP |
Room temperature (18-25 °C) | |
Silica Membrane Technology | |
Binding capacity: 7 μg | |
Genomic DNA | |
Isolation of DNA from forensic samples | |
1-3 μg (from buccal swabs) |
Safety and Handling
missing translation for 'shelfLife' : At least 1 year
- NucleoSpin DNA Forensic (10)
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H413-May cause long lasting harmful effects to aquatic life.
Supplemental information
- MIXTURE LIST-Contains : guanidium thiocyanate, 2-propanol, guanidium hydrochloride,ethanol, proteinasek