Learn More
Agilent BioTek Objectives for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
Brand: Agilent BioTek 1220545
This item is not returnable.
View return policy
Description
Select from a range of objectives that provide 2.5x to 60x imaging.
Specifications
Objective 60X, NA: .7 | |
Imaging Multi-Mode Reader |