missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI1 Mouse anti-Human, Clone: 3C12F9, Proteintech
Mouse Monoclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | ORAI1 |
---|---|
Clone | 3C12F9 |
Concentration | 1.02 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827905
|
Proteintech
66223-1-IG-20UL |
20 μL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16817905
|
Proteintech
66223-1-IG-150UL |
150 μL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.Specifications
ORAI1 | |
1.02 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
84876 | |
ORAI1 Fusion Protein Ag18050 | |
Primary | |
-20°C | |
ORAI1 |
3C12F9 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q96D31 | |
ORAI1 | |
IgG2a | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title