Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel System Replacement Parts
Replace UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel System Replacement Parts, for use with the EasyCast™ B1 Mini Gel System.
135.00 DKK - 715.00 DKK
Specifications
For Use With (Equipment) | Owl™ EasyCast B1 Mini Gel Electrophoresis System |
---|---|
Gel Size | Mini |
Product Line | Owl, EasyCast |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11932945
|
Thermo Scientific™
B1-CST |
Caster |
715.00 DKK
Each |
|
|||||
11942945
|
Thermo Scientific™
B1-GK |
Gasket |
135.00 DKK
Pack of 2 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
|||||
11952945
|
Thermo Scientific™
B1-UVT |
Gel Tray |
715.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
Replace your UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel System Replacement Parts, for use with the EasyCast™ B1 Mini Gel System.
For use with Owl EasyCast B1 Mini Gel Electrophoresis Systems.Specifications
Owl™ EasyCast B1 Mini Gel Electrophoresis System | |
Mini | |
Owl, EasyCast |