Learn More
Epredia™ Papanicolaou EA Stains
Control the color hues and intensities of your work with Epredia™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
Brand: Epredia™ 75511
View more versions of this product
Description
- EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
- EA-65 is used with OG-6 for non-gynecological staining.
- Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Specifications
EA-50 Papanicolaou Stain | |
EA-50; 3.79L (1 Gal.) | |
1 gal. |
Safety and Handling
- Papanicolaou Stain EA-50
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H370-Causes damage to organs.
- H371-May cause damage to organs.
Precautionary Statement
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, methanol, isopropanol, phosphotungstic acid, bismarck brown Y, eosin-Ydye, fast green fcf