Learn More
MilliporeSigma™ Papanicolaou's solution 1a Harris Hematoxylin Solution
For cytological cancer and cycle diagnosis
Brand:  MilliporeSigma™ 1.09253.0500
Encompass_Preferred
Product Code. 10741061
  • POA / 500mL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
  • Provides good nucleic staining in 2 to 3 min.
  • Nuclei are stained blue, dark violet to black
Specifications
Specifications
Plastic Bottle
1 L
15° to 25°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Papanicolaou's solution 1a Harris' hematoxylin solution for cytological cancer and cycle diagnosis
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethylene glycol
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.