Learn More
Masterflex™ PharMed™ BPT I/P™ Spooled Precision Pump Tubing
Protect your light-sensitive samples
11030.00 DKK - 13145.00 DKK
Specifications
Certifications/Compliance | FDA approved; Prop 65 NA (no Prop 65 chemicals) |
---|---|
Material | PharMed BPT |
Min. Operating Temperature (Metric) | -20°C |
Sterilization Method | Autoclave, Ethylene Oxide, Gamma Irradiation |
Max. Operating Temperature (Metric) | 100°C |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11715009
|
Masterflex™
95687-73 |
73 |
13145.00 DKK
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
11725009
|
Masterflex™
95687-82 |
82 |
11030.00 DKK
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
Beschreibung
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory Approvals to match your unique application
- Designed to withstand over 10,000 hours of tubing life, reducing downtime.
- Ideal for protecting sensitive fluids in biopharmaceutical applications and cell and tissue culture work, this tubing complies with USP class VI and FDA standards
- The opaque tan color is good for light-sensitive samples
- Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide, making it suitable for clean-in-place and steam-in-place cleaning and sterilization systems
- Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste
Spezifikation
FDA approved; Prop 65 NA (no Prop 65 chemicals) | |
-20°C | |
100°C |
PharMed BPT | |
Autoclave, Ethylene Oxide, Gamma Irradiation | |
Pump Tubing |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts