missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, 99.4%, MP Biomedicals™
1160.00 DKK
Chemical Identifiers
CAS | 5625-37-6 |
---|---|
Molecular Formula | C8H18N2O6S2 |
Molecular Weight (g/mol) | 302.36 |
InChI Key | IHPYMWDTONKSCO-UHFFFAOYSA-N |
Synonym | Piperazine-N, N'-bis[2-ethanesulfonic acid] |
PubChem CID | 79723 |
ChEBI | CHEBI:44933 |
IUPAC Name | 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
SMILES | C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
Description
Chemical Identifiers
5625-37-6 | |
302.36 | |
Piperazine-N, N'-bis[2-ethanesulfonic acid] | |
CHEBI:44933 | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
C8H18N2O6S2 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
79723 | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
Specifications
5625-37-6 | |
Crystalline Powder | |
C8H18N2O6S2 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid | |
79723 | |
302.4 |
White | |
0.994 | |
Piperazine-N, N'-bis[2-ethanesulfonic acid] | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O | |
302.36 | |
CHEBI:44933 | |
Pipes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title