Learn More
Cole-Parmer™ Indenti-plug™ Plastic Foam Stopper
Keep the contents of your containers secure with these durable and reusable stoppers. Indenti-plug™ Plastic Foam Stoppers can also be used as a cleaner for pipettes, tubes, vials and precision tubing.
675.00 DKK - 955.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Material | Plastic |
Product Type | Plug Stopper |
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
11735289
|
Cole-Parmer™
06298-80 |
45 to 50 mm |
955.00 DKK
Pack of 80 |
Estimated Shipment: 27-06-2025
Log in to see stock. |
|||||
11795299
|
Cole-Parmer™
06298-78 |
35 to 45 mm |
675.00 DKK
Pack of 100 |
|
|||||
Description
- Breathable, nonabsorbent, chemical resistant and heat tolerant
- Open-cell foam has a uniform density
- Leaves no fibers
- Autoclavable
Specifications
Autoclavable | |
White | |
Plastic | |
Plug Stopper |