Learn More
Buerkle™ General Purpose Funnels PP/PE
Designed for general purpose use. Buerkle™ Plastic Funnels feature a suspension eyelet for safe filling.
13.45 DKK - 36.92 DKK
Specifications
For Use With (Application) | For Foodstuffs |
---|---|
Includes | Suspension eyelet |
Product Type | Funnel |
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Material | Price | Quantity & Availability | |||
10659480
|
Buerkle™
9602-0040 |
40 mm | 5 mm | Polypropylene |
26.11 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
10347042
|
Buerkle™
9602-0060 |
60 mm | 8 mm | Polyethylene |
13.45 DKK
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||
10690733
|
Buerkle™
9602-0100 |
100 mm | 10 mm | Polypropylene |
26.11 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
10645873
|
Buerkle™
9602-0080 |
80 mm | 11 mm | Polyethylene |
26.11 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
10205772
|
Buerkle™
9602-0120 |
120 mm | 13 mm | Polypropylene |
26.11 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
10260032
|
Buerkle™
9602-0140 |
140 mm | 16 mm | Polyethylene |
30.22 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
10071722
|
Buerkle™
9602-0160 |
160 mm | 18 mm | Polyethylene |
36.92 DKK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||
Description
- Suitable for foodstuff
- Available in polyethylene or polypropylene
- Wide range of sizes
Specifications
For Foodstuffs | |
Suspension eyelet | |
Funnel |