Learn More
Buerkle™ Plastic Single-use Pipets
Pipettes made of plastic are particularly suited for training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry
25.35 DKK - 237.00 DKK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11832293
|
Buerkle™
7556-0050 |
50 mL |
53.20 DKK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
17210684
|
Buerkle™
7556-0025 |
25 mL |
25.35 DKK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
11842293
|
Buerkle™
7556-0100 |
100 mL |
237.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Polystyrene, crystal clear
- For aqueous solutions
- |Polystyrene, crystal clear
Specifications
Polystyrene | |
Sterile |