Learn More
Platinum Iridium wire, 1.5mm (0.06in) dia, Thermo Scientific Chemicals
10640.04 DKK - 42462.68 DKK
Specifications
Chemical Name or Material | Platinum Iridium wire |
---|---|
Name Note | 1.5mm (0.06 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 90:10 wt% |
MDL Number | MFCD02091683 |
Solubility Information | Insoluble in water and acid. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450101
|
Thermo Scientific Alfa Aesar
044812.BQ |
5 cm |
10640.04 DKK
5cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460101
|
Thermo Scientific Alfa Aesar
044812.BU |
25 cm |
42462.68 DKK
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum-iridium wire is used in platinum jewelry, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibers; in metrology and in fabrication of metal microelectrodes. It is used in the electrical stimulation of nervous tissue and in electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium wire | |
Pt:Ir; 90:10 wt% | |
Insoluble in water and acid. | |
≈1.91g/5cm |
1.5mm (0.06 in.) dia. | |
MFCD02091683 | |
Odorless | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures