Learn More
Memmert™ Plug-In Tube Extension for SN750plus, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ SN750plusB29718
343.20 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
60.3 mm | |
SN750plus Sterilizer |