Learn More
Macherey-Nagel™ Polyester Sheets Polygram Alox N/UV254 TLC Micro-Set M
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
4 x 8 cm
Unit Size:
Pack of 200
Description
TLC polyester sheets POLYGRAM Alox, aluminum oxide, neutral. We recommend to activate aluminum oxide layers before use.
Specifications
Specifications
Material | Polyester Sheets Polygram Alox N |
Quantity | 200 Pk. |
Type | TLC Micro-Set M |
Fluorescent Indicator | UV254 |
Includes | 2 developing chambers; 2 x 50 glass capillaries 1 μL; 1 rubber cap for glass capillaries; 2 spotting guides; 1 measuring cylinder 10 mL; 1 beaker 25 mL; 1 glass laboratory sprayer with rubber bulb; 1 plastic syringe 1 mL; 20 Sheets filter paper MN 713, 15 x 21 cm; 50 polyester sheets Polygram Alox N/UV254 |
Dimensions (L x W) | 4 x 8 cm |