Learn More
Bel-Art™ Scienceware™ Polyethylene Magnetic Stirring Bar Retriver
Eliminates contamination of contents Does not need to be cleaned or autoclaved after use Magnet is located in the 25 x 70mm rectangular section; overall length: 20cm (7.7 in.) Long ribbed handle provides a secure grip Made of high density polyethylene
Brand:  Bel-Art™ Scienceware™ F37775-0000
192.09 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Product Code. 11785064
  • 204.00 DKK / Each
Estimated Shipment: 10-06-2025
Explore more special offers
Description
Eliminates contamination of contents Does not need to be cleaned or autoclaved after use Magnet is located in the 25 x 70mm rectangular section; overall length: 20cm (7.7 in.) Long ribbed handle provides a secure grip Made of high density polyethylene
  • Eliminates contamination of contents
  • Does not need to be cleaned or autoclaved after use 
  • Magnet is located in the 25 x 70mm rectangular section; overall length: 20cm (7.7”)
  • Long ribbed handle provides a secure grip
  • Made of high density polyethylene
Specifications
Specifications
HDPE
Magnetic Stir Bar Retriever