Learn More
BioClean™ SafeStep NSO Cleanroom Overshoes
Overshoes have low levels of particle shedding and are processed to ensure ISO Class 5 compatibility. Elastomer coating ensures good grip and durability.
2820.00 DKK - 2985.00 DKK
Specifications
Material Polyethylene
Color White
Certifications/Compliance ISO 9001:2015
Height (Metric) 170 mm
Product Type Controlled Environments Shoe Covers
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
12952245
View Documents
BioClean™
NSO016
400 mm
2985.00 DKK
Pack of 400
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 05-12-2025
Add to basket
12962245
View Documents
BioClean™
NSO018
455 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Slip-resistant
  • Low levels of particle shedding
  • Durable
  • Elastomer coating
Specifications
Specifications
Polyethylene
ISO 9001:2015
Controlled Environments Shoe Covers
White
170 mm