Learn More
Sigma Laborzentrifugen™ Polypropylene Angle Rotors
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Polypropylene Angle Rotors are ideal for use in research laboratories.
293.00€ - 983.94€
Product Code | Brand | Capacity | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | For Use With (Equipment) | Price | Quantity & Availability | ||||
11720659
|
Sigma Laborzentrifugen™
12093 |
18 x 1.5/2 mL, 18 x 0.25/0.4 mL | Sigma 1-14 and 1-14K |
293.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
13425175
|
Sigma Laborzentrifugen™
12134 |
24 x 1.5/2 mL | Sigma 1-16 and 1-16K |
713.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
11780749
|
Sigma Laborzentrifugen™
12096 |
2 PCR Strips (8 x 0.2 mL) | Sigma 1-14 and 1-14K |
331.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10750594
|
Sigma Laborzentrifugen™
12094 |
24 x 1.5/2 mL | Sigma 1-14 and 1-14K |
387.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10566082
|
Sigma Laborzentrifugen™
12101 |
4 PCR Strips (8 x 0.2 mL) | Sigma 1-15, 1-15K, 1-15P, 1-15PK | N/A | |||||
Description
- Fixed-angle rotors are used for sedimentation with high centrifugal forces of up to and more than 60,000 x g
- Allow shorter sedimentation times in comparison to swing-bucket rotors
- Higher rotational speeds and higher centrifugal forces can be achieved
- Sedimentation distance is also generally shorter than with swing-bucket rotors
- Capacity of the rotor chamber can be better utilized than with swing-bucket rotors
- Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Polypropylene | |
Angle Rotor | |
Autoclavable |