Learn More
Tenak™ Polypropylene Cryobox, 80 mm Height
Cryobox with cell codes printed on the lid for easy identification.
1225.00 DKK - 1315.00 DKK
Specifications
Material | Polypropylene |
---|---|
Withstand Temperature (Metric) | As low as -90°C |
Dimensions (L x W x H) | 130 x 130 x 80 mm |
Description
- Constructed of polypropylene
- Can be used in temperatures as low as -90°C
- Autoclavable at 121°C for 20 minutes
- Boxes have a fixed grid divider with 9 x 9 cells and are suited for 1.2 mL and 2 mL cryogenic vials
Specifications
Polypropylene | |
As low as -90°C | |
130 x 130 x 80 mm |