Learn More
Brady™ Polypropylene: ISO Safety Sign - Direction arrow 90°, To the right, Green
Sign compliant to ISO 7010.
Brand: Brady™ 814422
This item is not returnable.
View return policy
Description
- Polypropylene
- For indoor and outdoor use
Specifications
ISO 7010 | |
Polypropylene | |
315 mm | |
315 mm | |
Square | |
White on Green |
PIC A090-315x315-PP-CRD/1 | |
Indoor and Outdoor | |
1.501 mm | |
-40°C to 120°C | |
Global |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu