Learn More
Cytiva Polypropylene Plunger
Brand: Cytiva 18-1031-80
352.70 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
XK 16 and AK 16 Columns | |
Plunger | |
1 Pack |
Polypropylene (Reinforced Glassfibre) | |
60°C |