Learn More
Porcelaines Avignon™ High Form Porcelain Crucibles
Heat and cool substances in the laboratory using Porcelaines Avignon™ Porcelain High Form Crucibles. They are available in various capacities.
Brand:  Porcelaines Avignon™ 11204
 View more versions of this product
Product Code. 10301124
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • High walls
  • Withstand high temperatures and rapid temperature changes
  • Porcelain
Specifications
Specifications
Porcelain
35mm
Crucible
17 mL
40mm