Learn More
Potassium cyanide, ACS Reagent, ≥96.0%, Honeywell Fluka™
Product Code. 15631790 Shop All Honeywell Fluka Products
2.5 kg, Poly Bottle
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
ACS Reagent,≥96.0%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]
Specifications
Melting Point 634°C (literature)
Color White
Physical Form Solid
Quantity 2.5 kg
Linear Formula KCN
UN Number UN1680
Beilstein 3593645
Solubility Information Soluble in water
Formula Weight 65.12g/mol
Percent Purity ≥96.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Granular Powder
Vapor Pressure 1.8 hPa at 634.5°C
Density 1.56g/cu.cm; Bulk Density: ca. 750kg/cu.m
Chemical Name or Material Potassium Cyanide
Show More Show Less
Product Content Correction
Product Title
Potassium cyanide, ACS Reagent, ≥96.0%, Honeywell Fluka™ >