Learn More
SPEX™ Prep-Aid Vertrel XF
SPEX™ Prep-Aid Vertrel XF is a liquid fluorocarbon grinding aid that prevents caking, reduces contamination, and evaporates after grinding.
Brand: SPEX™ 3650
This item is not returnable.
View return policy
Description
- Prevents caking and reduces contamination
- Quickly evaporates after grinding without leaving any residue
- Enhances the grindability of almost any sample
- Makes the grinding container easier to clean
- Particularly useful for grinding samples such as cement, rock, and clinker to particle sizes below 10 microns
Specifications
Grinding Aid | |
Grinding samples | |
946.35 mL bottle |