Learn More
Propper Steri-Wrap™ II Sterilization Wrap
Steri-Wrap™ II is a light weight cellulose based wrap designed for greater drapeability in steam and ethylene oxide sterilization cycles. Efficient microbial barrier, high-level of protection against fluids and particles. Highly permeable meaning no wet packs.
Brand:  Propper 11-890-9A
Encompass
 View more versions of this product
Product Code. 12307249
  • POA / Pack of 1000
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
  • Steri-Wrap™ is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process.
  • Steri-Wrap™ II is light weight wrap designed for great drapeability.
  • Steri-Wrap™ is designed to be virtually link free with tightly anchored fibers.
  • Wrap is reversible and features excellent liquid repellency.
  • Steri-Wrap™ can be used in steam or ethylene oxide sterilization.
  • Variety of strength of fabric to meet your needs.
Specifications
Specifications
Autoclavable
Single-use
Non-woven Fabric
12 x 12 in. (30.4 x 30.4 cm)
30.4 cm
30.4 cm
Blue, Green
Hospital, clinic and professional offices
Non-sterile
12 in.
12 in.
Steri-Wrap II
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.