Learn More
Lenz Laborglasinstrumente™ PTFE Ground Joint Sleeve with Grip Collar
202.00 DKK - 238.00 DKK
Specifications
Material | PTFE |
---|---|
Product Type | Ground Joint Sleeve |
Quantity | 1Pack |
Temperature Range (Metric) | –200 to 260°C |
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
10072983
|
Lenz Laborglasinstrumente™
01101719 |
NS 19/26 |
202.00 DKK
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10804541
|
Lenz Laborglasinstrumente™
01101729 |
NS 29/32 |
229.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
10894531
|
Lenz Laborglasinstrumente™
01101714 |
NS 14/23 |
238.00 DKK
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
Specifications
PTFE | |
Ground Joint Sleeve | |
1Pack | |
–200 to 260°C |