Learn More
Kartell™ Lining for Glass Tapers
Constructed of PTFE for high temperature and chemical resistance. Kartell™ Lining for Glass Tapers are specially designed to prevent machined glass joints from binding.
238.00 DKK - 380.00 DKK
Specifications
Certifications/Compliance | DIN 12242 |
---|---|
Material | PTFE |
For Use With | Glass Joint |
For Use With (Application) | Prevent glass joints from binding |
Packaging Quantity | 10/Pk. |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
10554851
|
Kartell™
0103800 |
14/23 Cone Size |
239.00 DKK
Pack of 10 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
10234851
|
Kartell™
0103900 |
19/26 Cone Size |
238.00 DKK
Pack of 10 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
10717741
|
Kartell™
0104100 |
29/32 Cone Size |
305.00 DKK
Pack of 10 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
10707741
|
Kartell™
0104000 |
24/29 Cone Size |
281.00 DKK
Pack of 10 |
Estimated Shipment: 23-09-2025
Log in to see stock. |
|||||
15845497
|
Kartell™
0104300 |
45/40 Cone Size |
380.00 DKK
Pack of 10 |
Estimated Shipment: 30-09-2025
Log in to see stock. |
|||||
Description
- Designed to prevent machined glass joints from binding
- Conforms to DIN 12242
- Suitable for foodstuff
Specifications
DIN 12242 | |
Glass Joint | |
10/Pk. |
PTFE | |
Prevent glass joints from binding |