Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Ensure long-lasting durability and prevent demagnetization of Vitlab™ PFTE Polygonal Magnetic Stirring Bars via proper storage and not dropping them on hard surfaces (e.g. steel).
Brand:  VITLAB™ 301097
 View more versions of this product
Product Code. 10366381
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Permanent magnet AlNiCo V core
  • Angled shape gives rise to significant turbulence
  • Achieves effective mixing, even at low spin rates
  • FTE
Specifications
Specifications
For preparation, separation and concentration
3 mm
PTFE
Polygon
Permanent magnet AlNiCo V core
10 mm
5/Pk.