Learn More
Pyridine-3-boronic acid, Thermo Scientific Chemicals
CAS: 1692-25-7 | C5H6BNO2 | 122.92 g/mol
488.11 DKK - 1467.30 DKK
Chemical Identifiers
CAS | 1692-25-7 |
---|---|
Molecular Formula | C5H6BNO2 |
Molecular Weight (g/mol) | 122.92 |
MDL Number | MFCD00674177 |
InChI Key | ABMYEXAYWZJVOV-UHFFFAOYSA-N |
Synonym | pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid |
PubChem CID | 2734378 |
IUPAC Name | pyridin-3-ylboronic acid |
SMILES | OB(O)C1=CC=CN=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338126
|
Thermo Scientific Alfa Aesar
L15040.03 |
1 g | N/A | ||||||
11348126
|
Thermo Scientific Alfa Aesar
L15040.06 |
5 g |
1467.30 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.
Solubility
Slightly soluble in methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
1692-25-7 | |
122.92 | |
ABMYEXAYWZJVOV-UHFFFAOYSA-N | |
2734378 | |
OB(O)C1=CC=CN=C1 |
C5H6BNO2 | |
MFCD00674177 | |
pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid | |
pyridin-3-ylboronic acid |
Specifications
1692-25-7 | |
C5H6BNO2 | |
471943 | |
Slightly soluble in methanol. | |
OB(O)C1=CC=CN=C1 | |
122.92 | |
122.92 |
>300°C | |
MFCD00674177 | |
pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid | |
ABMYEXAYWZJVOV-UHFFFAOYSA-N | |
pyridin-3-ylboronic acid | |
2734378 | |
Pyridine-3-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold