Learn More
Systec™ Quick Cooling Option for H-210, H-320 Autoclaves
Systec™ Quick Cooling Option for H-210, H-320 Autoclaves facilitates rapid, efficient, and safe cooling of autoclaved liquids.
Brand: Systec™ 6076
This item is not returnable.
View return policy
Description
- Uses cooling water coils surrounding the chamber to help rapidly cool the chamber down to a safe temperature.
Specifications
H-210, H-320 Autoclave | |
Water Cooling Coils Option |