Learn More
Tenak™ Eco Alurack for 20 mm Microtiterplates
Freezer rack designed to hold microtiterplates.
Brand:  Tenak™ TE27713
1640.59 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15250994
  • 1690.00 DKK / Each
Only null left
Estimated Shipment: 15-10-2025
Description
  • Rack
  • For use with freezers
Specifications
Specifications
Freezers
Microtiterplates (114)
19 x 6