Learn More
RBM4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | RBM4B |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16814574
|
Proteintech
15412-1-AP-20UL |
20 μL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16804574
|
Proteintech
15412-1-AP-150UL |
150 μL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play a role in alternative splice site selection during pre-mRNA processing.Specifications
RBM4B | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q64LC9, Q8VE92, Q9BQ04 | |
474154, 66704, 83759 | |
RBM4B Fusion Protein Ag7679 | |
Primary | |
-20°C | |
RBM4B |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RBM30, RBM4B, RBM4L, RNA binding motif protein 30, RNA binding motif protein 4B, RNA binding protein 30, RNA binding protein 4B, ZCCHC15, ZCRB3B | |
Rbm4b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |