Learn More
Medline Scientific™ Refractometer Abbe 240V 50Hz
Brand:  Medline Scientific™ 8300.0000M
2261.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11960459
  • 2325.00€ / Each
Currently Unavailable: We have no information when this product will be available.