Learn More
Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation
Ideal for routine laboratory analysis and procedures. Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation are manufactured from durable DURAN™ borosilicate glass.
330.00 DKK - 382.00 DKK
Specifications
Diameter (Metric) | 16 mm |
---|---|
Product Type | Glass Tube |
Material | Borosilicate Glass |
Height (Metric) | 100 mm |
Product Code | Brand | Closure Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Price | Quantity & Availability | |||||
10396412
|
Lenz Laborglasinstrumente™
05700007 |
Not Included |
382.00 DKK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10767571
|
Lenz Laborglasinstrumente™
05700107 |
Stopper |
330.00 DKK
Pack of 10 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||
Description
- With round bottom and NS socket
- Available without stopper, with PE stopper, or with hexagonal hollow glass stopper
- Corrosion-free and resistant to thermal shock
Specifications
16 mm | |
Glass Tube | |
Borosilicate Glass | |
100 mm |