Learn More
Greiner Bio-One Round Bottom Polypropylene Test Tubes with Screw Cap
Polypropylene displays high thermal, mechanical and chemical stability.
Brand: Greiner Bio-One 163261
This item is not returnable.
View return policy
Description
- Volume: 12mL
- White screw cap
- Dia. X Height: 16.8 x 100mm
- Available sterile and nonsterile
Specifications
White | |
100 mm | |
Sterile | |
Screw Cap |
12 mL | |
Polypropylene | |
16.8 mm | |
Test Tube |