missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPAP3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23741-1-AP-20UL
This item is not returnable.
View return policy
Description
The tetratricopeptide repeat (TPR) motif is a degenerate, 34 amino acid sequence found in many proteins and acts to mediate protein-protein interactions in various pathways. At the sequence level, there can be up to 16 tandem TPR repeats, each of which has a helix-turn-helix shape that stacks on other TPR repeats to achieve ligand binding specificity. RPAP3 (RNA polymerase II-associated protein 3) is a 665 amino acid protein that contains seven TRP repeats and is expressed as multiple alternatively spliced isoforms. Existing as a tightly associated component of the RNA polymerase II (Pol II) complex, RPAP3 functions to form an interface between Pol II and scaffolding proteins, thereby playing a role in the regulation of protein complex formation.Specifications
RPAP3 | |
Polyclonal | |
Unconjugated | |
RPAP3 | |
RPAP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79657 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H6T3 | |
RPAP3 | |
RPAP3 Fusion Protein Ag20661 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction