Learn More
Saint Gobain Life Sciences Tygon™ ND 100-65 Medical Tubing
Tygon™ ND 100-65 Medical Tubing for blood contact medical devices where non-DEHP plasticized materials are required.
136.00 DKK - 1950.00 DKK
Specifications
Material | Non-DEHP Plastic |
---|---|
Overall Length (English) | 50 ft. |
Overall Length (Metric) | 15.2 m |
Color | Clear |
Product Type | Tubing |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
10366933
|
Saint Gobain Life Sciences
ADF00002 |
1/16 in. | 1/8 in. | 1/32 in. |
374.00 DKK
15m |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
10328184
|
Saint Gobain Life Sciences
ADF00004 |
3/32 in. | 5/32 in. | 1/32 in. |
330.00 DKK
15m |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
12356839
|
Saint Gobain Life Sciences
ADF00012 |
3/16 in. | 5/16 in. | 1/16 in. |
749.00 DKK
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
10667925
|
Saint Gobain Life Sciences
ADF00017 |
1/4 in. | 0.375 in. | 1/16 in. |
892.00 DKK
15m |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
12396839
|
Saint Gobain Life Sciences
ADF00018 |
1/4 in. | 7/16 in. | 3/32 in. |
1480.00 DKK
15m |
Estimated Shipment: 04-11-2025
Log in to see stock. |
|||
12316849
|
Saint Gobain Life Sciences
ADF00022 |
5/16 in. | 7/16 in. | 1/16 in. |
1080.00 DKK
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
12306849
|
Saint Gobain Life Sciences
ADF00019-C |
1/4 in. | 0.5 in. | 1/8 in. |
1950.00 DKK
Each |
Estimated Shipment: 13-01-2026
Log in to see stock. |
|||
12346849
|
Saint Gobain Life Sciences
ADF00027 |
0.375 in. | 0.5 in. | 1/16 in. |
1240.00 DKK
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
12356849
|
Saint Gobain Life Sciences
ADF00028 |
0.375 in. | 0.563 in. | 3/32 in. |
1595.00 DKK
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
15692054
|
Saint Gobain Life Sciences
ADF00037 |
0.5 in. | 11/16 in. | 3/32 in. |
136.00 DKK
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||
Description
- Crystal clear for easy visualization of fluid flow
- For contact with bodily fluids
- Non-wetting surface permits complete drainage
- Non-hemolytic and non-pyrogenic
- Withstands EtO, gas and gamma sterilization
- Minimally invasive devices
- Blood and IV solutions
- Dialysis equipment
- Wound drainage
- Inhalation equipment
- Chemotherapy drug delivery
- Kidney dialysis pumps
- Peristaltic pumps
- Catheters
Compliance
Meets USP Class VI criteria, USP Class VI, ISO 10993, European Pharmacopoeia 3.1.1.2 and European REACH requirements.
Specifications
Non-DEHP Plastic | |
15.2 m | |
Tubing |
50 ft. | |
Clear |