Learn More
Sartorius Accessories for Combisart™ Systems: Woulff's Bottle
Bottle for use with Sartorius Combisart System
Brand: Sartorius 16610
1245.98 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Volume, 500mL
- With stopcock
- For protection of vacuum pump
- Used between suction flask and vacuum source
Specifications
Woulff's Bottle | |
500mL | |
Protection of vacuum pump, used between suction flask and vacuum source | |
Stop Cock |
None | |
Glass | |
Used between suction flask and vacuum source |