Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Brand: Thermo Scientific™ RH52-15
2010.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Centrifuge Tubes: 52 x 15L conical, Large capacity models | |
SPD210, SPD2030, SPD300DDA (requires RAD200) | |
RH52-15 | |
15 mL |
Rotor | |
52 15 mL Conical | |
Traditional |