Learn More
Scandium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
Specifications
Specifications
CAS | 7440-20-2 |
Molecular Formula | Sc |
MDL Number | MFCD00016323 |
InChI Key | SIXSYDAISGFNSX-UHFFFAOYSA-N |
SMILES | [Sc] |
IUPAC Name | scandium |
Solvent or Matrix | 2% HNO3 |
Molecular Weight (g/mol) | 44.96 |
PubChem CID | 23952 |
Quantity | 125 mL |