Learn More
SemperIt™ Semperguard™ Latex Innercoated (IC) Powder-Free Gloves
Manufactured from high quality soft latex film. Semperit™ Semperguard™ Latex Innercoated (IC) Powder-Free Gloves are available in lightly powdered and powder-free styles.
Brand:  SemperIt™ 3000001293
Product Code. 12980705
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Manufactured from high quality soft latex film and available in lightly powdered and powder-free styles
  • Lightly powdered or powder-free
  • Microroughened outer surface offeres better grip
  • Powder-free gloves have a synthetic inner coating for ease of donning
  • Ambidextrous 
  • Rolled cuff
Specifications
Specifications
CE Class I, EN 420, EN 374, EN 455, PPE Category III
Natural
Ambidextrous
Powder Free
Non-sterile
Suitable for food contact
90/Pk.
240 mm
Smooth inside, textured outside
Single-use
Rubber
X-Large
0.24 mm
Rolled
18/14 MPa