missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68173-1-IG-20UL
This item is not returnable.
View return policy
Description
The SEPT6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEPT6. This antibody recognizes human, mouse, pig, rabbit, rat antigen. The SEPT6 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
SEPT6 | |
Monoclonal | |
500 μg/mL | |
KIAA0128; MGC16619; MGC20339; SEP2; SEPT2 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human, Mouse, Pig, Rabbit, Rat | |
50 kDa | |
Liquid |
Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
1B1G1 | |
Unconjugated | |
Mouse | |
Protein G purification | |
RUO | |
23157 | |
Store at -20°C. Stable for one year after shipment. | |
Monoclonal Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction