Learn More
Trajan™ Diamond MS Autosampler Syringes: For Agilent
Product Code. p-4682046
Click to view available options
Volume (Metric):
10 μL
25 μL
Model:
10R-AG-GT-MS1
25R-AG-GT-MS1
Unit Size:
Each
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
For Use With (Application) Improves mass spectrometry analysis sensitivity
Product Type Diamond MS Syringe
Model 10R-AG-GT-MS1
Quantity Ea.
Volume (Metric) 10 μL
Connection Type Removable