Learn More
Trajan™ GC Capillary Column Connectors: Replacement μ-Unions
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
1150.00 DKK
Specifications
Type | Replacement μ-Union |
---|---|
For Use With (Equipment) | GC Capillary Column Connectors |
Product Code | Brand | Connections | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connections | Price | Quantity & Availability | |||||
15248943
|
Trajan™
073573 |
0.1 - 0.32 mm and 0.53 mm |
1150.00 DKK
Pack of 5 |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||||
15258943
|
Trajan™
073572 |
0.1 to 0.32 mm |
1150.00 DKK
Pack of 5 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||
15278943
|
Trajan™
073574 |
0.53 mm and 0.53 mm |
1150.00 DKK
Pack of 5 |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||||
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Replacement μ-Union | |
GC Capillary Column Connectors |