Learn More
Trajan™ GC Extended Stainless Steel Nuts
Provide optimum system connections for SGE fittings.
Brand: Trajan™ 103407
745.35 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with SGE fittings.
- Stainless Steel Construction
Specifications
Nut | |
Stainless Steel | |
1/4 in. Hole (Extended) |